Learn More
Discovery Life Sciences UGT REACTION MIX A
SDP
Catalog No. NC9005071
Encompass
UGT Reaction Mix Solution A, 2mL, Store at -20degreeC, Components:25mM Uridine 5'-Diphospho-Glucuronic Acid (UDPGA) in water, This product showed no loss in performance after 10 freeze/thaw cycles.
Catalog No. NC9005071 Supplier Discovery Life Sciences Supplier No. 451300
Add to Cart
Add to Cart
UGT Reaction Mix Solution A, 2mL, Store at -20degreeC, Components:25mM Uridine 5'-Diphospho-Glucuronic Acid (UDPGA) in water, This product showed no loss in performance after 10 freeze/thaw cycles.