Learn More
Aviva Systems Biology Corp UGT1A1 Antibody - N-terminal region (ARP44292_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44292_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against UGT1A1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human UGT1A1. Alternate names: GNT1, HUG-BR1, UDPGT, UGT1, UGT1A, BILIQTL1, UDPGT 1-1ApplicationWB. Species reactivity: Horse, Human, Mouse, Rat
Catalog No. 50-014-58773