Learn More
Aviva Systems Biology Corp UGT1A4 Antibody - N-terminal region (ARP46829_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46829_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against UGT1A4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human UGT1A4. Alternate names: HUG-BR2, UDPGT, UGT1D, UGT-1D, UGT1.4, UGT1-04, UDPGT 1-4ApplicationWB. Species reactivity: Human
Catalog No. 50-014-59413