Learn More
Aviva Systems Biology Corp UGT1A5 Antibody - N-terminal region (ARP49439_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49439_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against UGT1A5. It was validated on Western Blot. UGT1A5,UDP glucuronosyltransferase 1 family, polypeptide A5,UDPGT; UGT1E; UDPGT 1-5; UGT1A5,UDP-glucuronosyltransferase 1-5,UDP-glucuronosyltransferase 1-5,UGT-1E,UGT1*5,OTTHUMP00000065199,UDP-glucuronosyltransferase 1-E,UDP-glucuronosyltransferase 1A5,UDP glycosyltransferase 1 family, polypeptide A5,UDP-glucuronosyltransferase 1 family polypeptide A5sThe immunogen is a synthetic peptide directed towards the N terminal region of human UGT1A5. Alternate names: UDPGT, UGT1E, UDPGT 1-5ApplicationWB. Species reactivity: Human, Rat
Catalog No. 50-285-419