Learn More
Aviva Systems Biology Corp UGT1A6 Antibody - C-terminal region (ARP41458_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41458_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against UGT1A6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human UGT1A6. Alternate names: GNT1, HLUGP, HLUGP1, MGC29860, UDPGT, UGT1, UGT1F, UGT1A6S, UDPGT 1-6ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-57866