Learn More
Aviva Systems Biology Corp UGT1A7 Antibody - N-terminal region (ARP49438_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49438_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against UGT1A7. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human UGT1A7. Alternate names: UDPGT, UGT1G, UGT-1G, UGT1.7, UGT1-07, UDPGT 1-7ApplicationIHC, WB. Species reactivity: Human
Catalog No. 50-014-60086