Learn More
Aviva Systems Biology Corp UGT2A3 Antibody - N-terminal region (ARP49861_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49861_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against UGT2A3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human UGT2A3. Alternate names: FLJ21934ApplicationWB. Species reactivity: Human
Catalog No. 50-014-60203