Learn More
Aviva Systems Biology Corp UGT2B15 Antibody - N-terminal region (ARP41906_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41906_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against UGT2B15. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human UGT2B15. Alternate names: UGT2B8, HLUG4, UDPGTH3, UDPGT 2B8, UDPGT2B15ApplicationWB. Species reactivity: Human
Catalog No. 50-014-53318