Learn More
Aviva Systems Biology Corp UGT3A2 Antibody - middle region (ARP49120_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49120_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against UGT3A2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human UGT3A2. Alternate names: MGC119426, MGC119429ApplicationWB. Species reactivity: Dog, Human
Catalog No. 50-014-53573