Learn More
Cayman Chemical UGT8 Inhibitor 19, Purity: ≥98%, CAS: 2414349-93-0, 1 mg
SDP
Catalog No. 502947088
Encompass_Preferred
An inhibitor of UGT8 (IC50 = 0.2 nM); selective for UGT8 over UGT1A1, -1A6, -2B7, -2B15, and -2B17 (IC50s = >10 µM for all); inhibits the synthesis of GalCer and SFT in juvenile mouse brain (ED50s = <3 mg/kg for both) and adult mouse kidney at 3, 10, and 30 mg/kg
Catalog No. 50-294-7088 Supplier Cayman Chemical Supplier No. 327231 MG
An inhibitor of UGT8 (IC50 = 0.2 nM); selective for UGT8 over UGT1A1, -1A6, -2B7, -2B15, and -2B17 (IC50s = >10 µM for all); inhibits the synthesis of GalCer and SFT in juvenile mouse brain (ED50s = <3 mg/kg for both) and adult mouse kidney at 3, 10, and 30 mg/kg