Learn More
Aviva Systems Biology Corp Uhmk1 Antibody - C-terminal region (ARP41180_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41180_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Uhmk1. It was validated on Western Blot. . Alternate names: 4732477C12Rik, 4930500M09Rik, AA673513, AI449218, AU021979, KIS, KistApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-284-917