Learn More
IDEX Health & Science Upchurch UHP MICROTIGHT UNION
Catalog No. NC0771921
Encompass_Preferred
Our VHP MicroTight Union features a durable stainless steel body and a unique stainless steel union capsule. This enables excellent chemical compatibility and conductivity making it an ideal choice for electrical interfacing in certain LC-MS applications. It is for 360um OD tubing and includes (2) PK-152 ferrules and (2) P-416BLK nuts.
Catalog No. NC0771921 Supplier IDEX Health & Science Upchurch Supplier No. UH436
Our VHP MicroTight Union features a durable stainless steel body and a unique stainless steel union capsule. This enables excellent chemical compatibility and conductivity making it an ideal choice for electrical interfacing in certain LC-MS applications. It is for 360um OD tubing and includes (2) PK-152 ferrules and (2) P-416BLK nuts.