Learn More
Perkin Elmer US LLC UHP-35ZA-S Zero Air Generator - 15 L/min, 230 V
Catalog No. 502112878
Encompass_Preferred
These generators produce zero-grade air with less than 0.05 ppm hydrocarbon content, measured as methane, enabling a more stable baseline as compared to conventional sources of fuel air.
Catalog No. 50-211-2878 Supplier Perkin Elmer US LLC Supplier No. N9303209
Add to Cart
Add to Cart
Request a Quote
These generators produce zero-grade air with less than 0.05 ppm hydrocarbon content, measured as methane, enabling a more stable baseline as compared to conventional sources of fuel air.