Learn More
WELCH MATERIALS INC Ultisil UHPLC AQ-C18, 1.8µm, 2.1×50mm
Catalog No. NC2052547
Encompass_Preferred
Ultisil UHPLC AQ-C18, 1.8μm, 2.1×50mm, pH range: 1.5-10.0, Pore Size: 120Å, USP listing: L1, Carbon Loading: 12%, Surface Area: 320m2/g, endcapped.
Catalog No. NC2052547 Supplier WELCH MATERIALS INC Supplier No. H0020711010
Add to Cart
Add to Cart
Ultisil UHPLC AQ-C18, 1.8μm, 2.1×50mm, pH range: 1.5-10.0, Pore Size: 120Å, USP listing: L1, Carbon Loading: 12%, Surface Area: 320m2/g, endcapped.