Learn More
Perkin Elmer US LLC UHPLC – General Starter Kit
Supplier: Perkin Elmer US LLC N2930801
This kit contains: Open-End Wrench Adjustable Wrench Reversible Screw Driver Parker-Hannifin ZDV Stainless Steel Union Parker-Hannifin Medium-Stem Stainless Steel Nuts 116 in. Parker-Hannifin Stainless Steel Ferrules 116 in. Valco Stainless Steel Ferrules Finger-Tight Fittings, PEEK, 5.5K psi Max (pkg. 5) Optimize Technologies EZP (Hand-Tight Fitting to 13K psi, Wrench-tight to 20K psi) 116 in. Tefzel Nuts, 116 in. Tefzel Reverse Ferrules 18 in. Tefzel Nuts 4 N2601189 18 in. Tefzel Reverse Ferrules 4 09903771 PEEK Tubing Cutter (Includes 5 Extra Blades) 1 N9307820 50 cm x 0.005 in. ID 116 in. Stainless Steel Tubing 3 02507060 5 ft. x 0.004 in. ID 116 in. PEEK Tubing, Black 1 N2916261 5 ft. x 0.005 in. ID 116 in. PEEK Tubing, Red 1 N2916262 30 mL Priming Syringe 1 09904849 2 mL Clear Write-On Vials w/ 10 mm Pre-Slit PTFE/Silicone Caps (pkg. 100) 1 N9300695 Cable/Tubing Organizer Clip