Learn More
WELCH MATERIALS INC Ultisil UHPLC LP-C18 1.8µm, 2.1×50mm W-PORT
Catalog No. NC2052553
Encompass_Preferred
Ultisil UHPLC LP-C18, 1.8μm, 2.1×50mm, W-Port, pH range: 0.5-8.0, Pore Size: 120Å, USP listing: L1, Carbon Loading: 10%, Surface Area: 320m2/g, not-endcapped.
Catalog No. NC2052553 Supplier WELCH MATERIALS INC Supplier No. H0020811210
Ultisil UHPLC LP-C18, 1.8μm, 2.1×50mm, W-Port, pH range: 0.5-8.0, Pore Size: 120Å, USP listing: L1, Carbon Loading: 10%, Surface Area: 320m2/g, not-endcapped.