Learn More
WELCH MATERIALS INC Ultisil UHPLC POLAR RP,1.8µm,2.1×50mm
Catalog No. NC2052560
Encompass_Preferred
Ultisil UHPLC Polar RP, 1.8μm, 2.1×50mm, pH range: 1.5-10.0, Pore Size: 120Å, USP listing: L1, L60, Carbon Loading: 18%, Surface Area: 320m2/g, endcapped.
Catalog No. NC2052560 Supplier WELCH MATERIALS INC Supplier No. H0021511010
Ultisil UHPLC Polar RP, 1.8μm, 2.1×50mm, pH range: 1.5-10.0, Pore Size: 120Å, USP listing: L1, L60, Carbon Loading: 18%, Surface Area: 320m2/g, endcapped.