Learn More
Thermo Scientific Liquid Junction Column Outlet IDXODXL 20X3 [1]
Catalog No. 501347912 Shop All Thermo Scientific Products
Encompass
1m x 360μm x 20μm fused silica tubing with polyimide coating column outlet refractive index detector for connecting liquid chromatography columns to mass spectrometry systems with low dispersion and minimal peak broadening withstands pressures up to 580 bar.
Catalog No. 50-134-7912 Supplier Thermo Scientific Supplier No. 6041.5290
1m x 360μm x 20μm fused silica tubing with polyimide coating column outlet refractive index detector for connecting liquid chromatography columns to mass spectrometry systems with low dispersion and minimal peak broadening withstands pressures up to 580 bar.