Learn More
WELCH MATERIALS INC Ultisil UHPLC XB-C18, 1.8µm, 2.1×50mm
Catalog No. NC2052543
Encompass_Preferred
Ultisil UHPLC XB-C18, 1.8μm, 2.1×50mm, pH range: 1.5-10.0, Pore Size: 120Å, USP listing: L1, Carbon Loading: 17%, Surface Area: 320m2/g, endcapped.
Catalog No. NC2052543 Supplier WELCH MATERIALS INC Supplier No. H0020111010
Ultisil UHPLC XB-C18, 1.8μm, 2.1×50mm, pH range: 1.5-10.0, Pore Size: 120Å, USP listing: L1, Carbon Loading: 17%, Surface Area: 320m2/g, endcapped.