Learn More
Mac-Mod Fortis SpeedCore BIO INERT Peptide C18, 160 Å, 2.6 µm, 2.1 x 50 mm Core Shell Column in Inert-Coated Hardware
SDP
Catalog No. 5000151615
Encompass
Fortis SpeedCore Core-Shell columns are the very latest in Core-Shell technology. Incorporating optimized bonding density and unique stringent packing processes, the new 80 A Core-Shell columns provide the chromatographer with the ability to speed up analysis and increase resolution over traditional fully porous 3 and 5 micron materials. Fortis SpeedCore is available in a wide range of bonded phase chemistries including C18, C18 pH+, C18-PFP, C8, Diphenyl, RP18-Amide, AQUA, and PFP bonded phases and 2.6 and 5 um particle sizes. Fortis SpeedCore columns are available in analytical formats.
Catalog No. 50-001-51615 Supplier Mac-Mod Supplier No. SCPEP18020326M
Fortis SpeedCore Core-Shell columns are the very latest in Core-Shell technology. Incorporating optimized bonding density and unique stringent packing processes, the new 80 A Core-Shell columns provide the chromatographer with the ability to speed up analysis and increase resolution over traditional fully porous 3 and 5 micron materials. Fortis SpeedCore is available in a wide range of bonded phase chemistries including C18, C18 pH+, C18-PFP, C8, Diphenyl, RP18-Amide, AQUA, and PFP bonded phases and 2.6 and 5 um particle sizes. Fortis SpeedCore columns are available in analytical formats.