Learn More
Mac-Mod Fortis SpeedCore INERT C18-PFP, 80 Å, 2.6 µm, 2.1 x 50 mm Core Shell Column in Inert-Coated Hardware
SDP
Catalog No. 5000152225
Encompass
Fortis SpeedCore HILIC Core-Shell columns are the latest in Core-Shell technology. Incorporating optimized bonding density and unique stringent packing processes, the new 80 A Core-Shell columns provide the chromatographer with the ability to speed up analysis and increase resolution over traditional fully porous 3 and 5 micron materials. Fortis SpeedCore HILIC is available in 2.6 and 5 um particle sizes. Fortis SpeedCore HILIC columns are available in analytical formats.
Catalog No. 50-001-52225 Supplier Mac-Mod Supplier No. SP18FP020326M
Fortis SpeedCore HILIC Core-Shell columns are the latest in Core-Shell technology. Incorporating optimized bonding density and unique stringent packing processes, the new 80 A Core-Shell columns provide the chromatographer with the ability to speed up analysis and increase resolution over traditional fully porous 3 and 5 micron materials. Fortis SpeedCore HILIC is available in 2.6 and 5 um particle sizes. Fortis SpeedCore HILIC columns are available in analytical formats.