Learn More
Aviva Systems Biology Corp UHRF2 Antibody - N-terminal region (ARP34772_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34772_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against UHRF2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human UHRF2. Alternate names: NIRF, URF2, RNF107, RP11-472F14.2ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55556