Learn More
Aviva Systems Biology Corp UHRF2 Antibody - middle region (ARP34620_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34620_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against UHRF2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human UHRF2. Alternate names: DKFZp434B0920, DKFZp686G0837, MGC33463, NIRF, RNF107, RP11-472F14.2, URF2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55483