Learn More
All Foils Ultra High Vacuum (UHV) Aluminum Foil, .001" X 18" X 500 LF
SDP
Catalog No. NC1701139
Encompass
This item has a minimum order quantity of 2 rolls per supplier requirements. For many of our customers, clean aluminum foil is not only a wish, it is a necessity. That is why All Foils, Inc. offers a full line of Ultra High Vacuum (UHV) Foil. Clean aluminum foil is virtually contamination free. This means the foil is “A” wettable and completely free from oils normally found on other grades of aluminum foil. Our UHV foil is produced on plastic cores. Each UHV Roll is also enclosed in a free plastic dispenser box and wrapped in film. This is important for customers who require that little or no contamination infiltrates their environment.
Catalog No. NC1701139 Supplier All Foils Supplier No. .001 X 18 X 500 LF
This item has a minimum order quantity of 2 rolls per supplier requirements. For many of our customers, clean aluminum foil is not only a wish, it is a necessity. That is why All Foils, Inc. offers a full line of Ultra High Vacuum (UHV) Foil. Clean aluminum foil is virtually contamination free. This means the foil is “A” wettable and completely free from oils normally found on other grades of aluminum foil. Our UHV foil is produced on plastic cores. Each UHV Roll is also enclosed in a free plastic dispenser box and wrapped in film. This is important for customers who require that little or no contamination infiltrates their environment.