Learn More
Strem, An Ascensus Company CAS# 1072413-89-8. 2g. Zirconium 1,4-dicarboxybenzene MOF (UiO-66).
SDP
Supplier:  Strem, An Ascensus Company 4011052G
Encompass
CAS# 1072413-89-8. 2g. Zirconium 1,4-dicarboxybenzene MOF (UiO-66). . Molecular Weight: 1664.06. Molecular Formula: C48H28O32Zr6. Color/form: white pwdr. Strem# 40-1105. http://www.strem.com/catalog/v/40-1105/
Catalog No. 50-204-5304