Learn More
AdipoGen UK-1
SDP
Supplier:  AdipoGen BVT0013M001
Encompass_Preferred
Chemical. CAS 151271-53-3. Formula C22H14N2O5. MW 386.4. Isolated from Streptomyces sp. K17/9. Antibiotic. Antifungal. Topoisomerase II Topo II inhibitor. Mg2- and Zn2-dependent DNA binding agent. Potent anticancer compound.
Catalog No. 50-256-2148