Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC UK-101 | 1000313-40-5 | 99.6% | 484.74 | C25H48N2O5Si | 1 MG
SDP

Catalog No. 5000307510
Encompass_Preferred

UK-101 is a selective small-molecule inhibitor of the immunoproteasome β1i (LMP2) subunit used in biochemical and cellular research to probe immunoproteasome function and induce apoptosis in certain cancer models.

  • Selective LMP2 (β1i) inhibitor with reported IC50 of 104 nM.
  • Shows ~144-fold selectivity versus β1c and ~10-fold versus β5.
  • High reported purity (99.58%) suitable for research applications.
  • Molecular weight 484.74 g·mol⁻¹ and formula C25H48N2O5Si.
  • Soluble in DMSO and supplied in small research quantities, including 1 mg packs.

Catalog No. 50-003-07510 Supplier Medchemexpress LLC Supplier No. HY1190371MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

UK-101 is a selective small-molecule inhibitor of the immunoproteasome β1i (LMP2) subunit used in biochemical and cellular research to probe immunoproteasome function and induce apoptosis in certain cancer models.

  • Selective LMP2 (β1i) inhibitor with reported IC50 of 104 nM.
  • Shows ~144-fold selectivity versus β1c and ~10-fold versus β5.
  • High reported purity (99.58%) suitable for research applications.
  • Molecular weight 484.74 g·mol⁻¹ and formula C25H48N2O5Si.
  • Soluble in DMSO and supplied in small research quantities, including 1 mg packs.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.