Learn More
Medchemexpress LLC Uk-101 | 1000313-40-5 | 99.6% | 484.74 | C25H48N2O5Si | 10 MG
SDP
Catalog No. 5000307509
Encompass_Preferred
UK-101 is a potent, selective inhibitor of the immunoproteasome β1i (LMP2) subunit used as a biochemical probe to study immunoproteasome activity and apoptosis-related pathways.
  • Potent LMP2 inhibition (IC50 ≈ 104 nM).
  • High selectivity versus β1c and β5 subunits.
  • High purity suitable for research use (≈99.6%).
  • Soluble in DMSO (≈7.3 mg/mL); may require warming or sonication.
  • Supplied in small quantities appropriate for laboratory assays.
  • Commonly used in biochemical assays and apoptosis research.
Catalog No. 50-003-07509 Supplier Medchemexpress LLC Supplier No. HY11903710MG
UK-101 is a potent, selective inhibitor of the immunoproteasome β1i (LMP2) subunit used as a biochemical probe to study immunoproteasome activity and apoptosis-related pathways.
  • Potent LMP2 inhibition (IC50 ≈ 104 nM).
  • High selectivity versus β1c and β5 subunits.
  • High purity suitable for research use (≈99.6%).
  • Soluble in DMSO (≈7.3 mg/mL); may require warming or sonication.
  • Supplied in small quantities appropriate for laboratory assays.
  • Commonly used in biochemical assays and apoptosis research.