Learn More
Medchemexpress LLC UK-101 | 1000313-40-5 | 99.6% | 484.74 g/mol | C25H48N2O5Si | 25 MG
SDP
Catalog No. 5000307511
Encompass_Preferred
UK-101 is a potent, selective immunoproteasome β1i (LMP2) inhibitor intended for research use. It has a reported IC50 of 104 nM against LMP2, with significant selectivity over the constitutive β1c and β5 proteasome subunits. UK-101 is used in cell-based studies to induce apoptosis and to investigate immunoproteasome function and cancer-related pathways. Supplied as a powder with recommended storage conditions to preserve stability.
  • Potent LMP2 (β1i) inhibitor with IC50 ≈ 104 nM.
  • Selective over proteasome β1c and β5 subunits.
  • Induces apoptosis in cell-based assays.
  • High purity suitable for biochemical and cellular studies.
  • Supplied as powder with recommended storage for long-term stability.
Catalog No. 50-003-07511 Supplier Medchemexpress LLC Supplier No. HY11903725MG
UK-101 is a potent, selective immunoproteasome β1i (LMP2) inhibitor intended for research use. It has a reported IC50 of 104 nM against LMP2, with significant selectivity over the constitutive β1c and β5 proteasome subunits. UK-101 is used in cell-based studies to induce apoptosis and to investigate immunoproteasome function and cancer-related pathways. Supplied as a powder with recommended storage conditions to preserve stability.
  • Potent LMP2 (β1i) inhibitor with IC50 ≈ 104 nM.
  • Selective over proteasome β1c and β5 subunits.
  • Induces apoptosis in cell-based assays.
  • High purity suitable for biochemical and cellular studies.
  • Supplied as powder with recommended storage for long-term stability.