Learn More
TARGETMOL CHEMICALS INC UK-240455 5MG
SDP
Catalog No. 502581365
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. UK-240455 is a potent and selective N-methyl-D-aspartic acid (NMDA) glycine receptor antagonist with neuroprotective effects and improved motor function in Parkinson's disease models and is a potential candidate compound for treatment of Parkinson's disease. purity: 99%
Catalog No. 50-258-1365 Supplier TARGETMOL CHEMICALS INC Supplier No. T132485MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. UK-240455 is a potent and selective N-methyl-D-aspartic acid (NMDA) glycine receptor antagonist with neuroprotective effects and improved motor function in Parkinson's disease models and is a potential candidate compound for treatment of Parkinson's disease. purity: 99%