Learn More
Medchemexpress LLC HY-107394 1mg Medchemexpress, UK 356618 CAS:230961-08-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1073941MG
Encompass_Preferred
Medchemexpress, HY-107394 1mg UK 356618 CAS:230961-08-7 UK 356618 (Compound 4j) is a potent and selective inhibitor of matrix metalloprotease-3 (MMP-3) with an IC50 of 5.9 nM. UK 356618 is less potent against MMP-1, MMP-2, MMP-9, MMP-13 and MMP-14 compared with MMP-3. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8402