Learn More
TARGETMOL CHEMICALS INC UK-383367 5MG
SDP
Catalog No. 502579598
Encompass_Preferred
Also available in 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. UK-383367 is a procollagen C-proteinase inhibitor with IC50 of 44 nM has excellent selectivity over MMPs. purity: 100%
Catalog No. 50-257-9598 Supplier TARGETMOL CHEMICALS INC Supplier No. T60645MG
Also available in 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. UK-383367 is a procollagen C-proteinase inhibitor with IC50 of 44 nM has excellent selectivity over MMPs. purity: 100%