Learn More
Selleck Chemical LLC UK 383367 10mg 348622-88-8
SDP
Catalog No. 505740
Encompass_Preferred
UK-383367 is a procollagen C-proteinase inhibitor with IC50 of 44 nM, has excellent selectivity over MMPs. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-574-0 Supplier Selleck Chemical LLC Supplier No. S222410MG
Add to Cart
Add to Cart
UK-383367 is a procollagen C-proteinase inhibitor with IC50 of 44 nM, has excellent selectivity over MMPs. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.