Learn More
Medchemexpress LLC HY-15475 10mg , UK-5099 CAS:56396-35-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1547510MG
Encompass_Preferred
HY-15475 10mg UK-5099 CAS: 56396-35-1 UK-5099 (PF-1005023) is a potent inhibitor of the mitochondrial pyruvate carrier (MPC). UK-5099 (PF-1005023) inhibits pyruvate-dependent O2 consumption with an IC50 of 50 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0165