Learn More
Selleck Chemical LLC UK 5099 10mg 56396-35-1
SDP
Supplier:  Selleck Chemical LLC S531710MG
Encompass_Preferred
UK5099 is a potent inhibitor of the mitochondrial pyruvate carrier, inhibiting pyruvate transport across the plasma membrane of trypanosomes with Ki value of 49 ?M. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8140