Learn More
Medchemexpress LLC UK-78282 hydrochloride | 136647-02-4 | 99.82% | 466.05 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYW0137275MG
Encompass_Preferred
UK-78282 is a novel piperidine, potent and selective Kv1.3 blocker with an IC50 of 200 nM. It effectively suppresses human T-lymphocyte activation in vitro. UK-78,282 binds to residues at the inner surface of the channel, overlapping the site of action of verapamil.
  • IC50 & Target: 200 nM (Kv1.3)
  • Appearance: Solid
  • Color: White to off-white
Catalog No. 50-004-54223