Learn More
UK Biobank Axiom™ Array Kit
Specifications
Specifications
| Type | Array Kit |
| Array | Genotyping |
| Number of Arrays | 96 arrays |
| Format | 96-array Plate |
| Species | Human |
| Content And Storage | • 1x96-format array plate. Store at 2–8°C. • 1 Axiom 2.0 Reagent Kit (includes reagents and controls for 1x96-format array plate). Storage can vary for each reagent; refer to product insert. • 1 Axiom GeneTitan Consumables Kit (includes GeneTitan instrument consumables to process 1x96 array plate. Store at room temperature. |
| For Use With (Application) | Microarray Analysis |
| Product Line | Axiom |
| Quantity | 1 each |
| Shipping Condition | Approved for shipment at Room Temperature or on Wet or Dry Ice |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.