Learn More
Medchemexpress LLC Uk122 | 940290-58-4 | 99.9% | 291.30 g/mol | C17H13N3O2 | 5 MG
SDP
Catalog No. 5000264792
Encompass_Preferred
A selective small-molecule inhibitor of urokinase-type plasminogen activator (uPA) for biochemical and cellular research into protease-mediated processes such as cancer cell migration and invasion. It is a 4-oxazolidinone analogue with reported uPA inhibition (IC50 0.2 μM). Supplied as a research reagent, not for human or veterinary use.
  • Selective uPA inhibition with IC50 of 0.2 μM.
  • 4-oxazolidinone analogue suitable for mechanistic studies.
  • High purity (~99.9%) for reproducible biochemical assays.
  • Small-molecule format compatible with cell-based and enzymatic assays.
  • Available in small research quantities for laboratory use.
Catalog No. 50-002-64792 Supplier Medchemexpress LLC Supplier No. HY1110565MG
A selective small-molecule inhibitor of urokinase-type plasminogen activator (uPA) for biochemical and cellular research into protease-mediated processes such as cancer cell migration and invasion. It is a 4-oxazolidinone analogue with reported uPA inhibition (IC50 0.2 μM). Supplied as a research reagent, not for human or veterinary use.
  • Selective uPA inhibition with IC50 of 0.2 μM.
  • 4-oxazolidinone analogue suitable for mechanistic studies.
  • High purity (~99.9%) for reproducible biochemical assays.
  • Small-molecule format compatible with cell-based and enzymatic assays.
  • Available in small research quantities for laboratory use.