Learn More
Medchemexpress LLC UK122 TFA | 940290-58-4 | 99.85% | 405.33 | 1 ML
SDP
Catalog No. 5000343763
Encompass_Preferred
UK122 is a potent and selective urokinase-type plasminogen activator (uPA) inhibitor with an IC50 of 0.2 μM. It exhibits little to no inhibition of tissue-type PA (tPA), plasmin, thrombin, and trypsin (all IC50>100 μM). This 4-oxazolidinone analogue acts as an anticancer agent by inhibiting cancer cell migration and invasion.
  • Potent and selective urokinase-type plasminogen activator (uPA) inhibitor.
  • IC50 of 0.2 μM for uPA.
  • Minimal inhibition of tPA, plasmin, thrombin, and trypsin (IC50 > 100 μM).
  • 4-oxazolidinone analogue.
  • Acts as an anticancer agent.
  • Inhibits cancer cell migration and invasion.
  • For research use only.
Catalog No. 50-003-43763 Supplier Medchemexpress LLC Supplier No. HY11105610MG
UK122 is a potent and selective urokinase-type plasminogen activator (uPA) inhibitor with an IC50 of 0.2 μM. It exhibits little to no inhibition of tissue-type PA (tPA), plasmin, thrombin, and trypsin (all IC50>100 μM). This 4-oxazolidinone analogue acts as an anticancer agent by inhibiting cancer cell migration and invasion.
  • Potent and selective urokinase-type plasminogen activator (uPA) inhibitor.
  • IC50 of 0.2 μM for uPA.
  • Minimal inhibition of tPA, plasmin, thrombin, and trypsin (IC50 > 100 μM).
  • 4-oxazolidinone analogue.
  • Acts as an anticancer agent.
  • Inhibits cancer cell migration and invasion.
  • For research use only.