Learn More
Medchemexpress LLC UK122 TFA | 940290-58-4 | 99.85% | 405.33 | 100 MG
SDP
Catalog No. 5000343247
Encompass_Preferred
UK122 TFA is a potent and selective urokinase-type plasminogen activator (uPA) inhibitor with an IC50 of 0.2 μM. It exhibits minimal or no inhibition of tissue-type PA (tPA), plasmin, thrombin, and trypsin (all IC50 >100 μM). This 4-oxazolidinone analogue acts as an anticancer agent that hinders cancer cell migration and invasion. This product is intended for research use only.
  • Potent and selective uPA inhibitor
  • Inhibits cancer cell migration and invasion
  • Minimal inhibition of tPA, plasmin, thrombin, and trypsin
  • Suitable for in vitro studies
Catalog No. 50-003-43247 Supplier Medchemexpress LLC Supplier No. HY11105650MG
UK122 TFA is a potent and selective urokinase-type plasminogen activator (uPA) inhibitor with an IC50 of 0.2 μM. It exhibits minimal or no inhibition of tissue-type PA (tPA), plasmin, thrombin, and trypsin (all IC50 >100 μM). This 4-oxazolidinone analogue acts as an anticancer agent that hinders cancer cell migration and invasion. This product is intended for research use only.
  • Potent and selective uPA inhibitor
  • Inhibits cancer cell migration and invasion
  • Minimal inhibition of tPA, plasmin, thrombin, and trypsin
  • Suitable for in vitro studies