Learn More
Medchemexpress LLC UK122 TFA | 940290-58-4 | 99.85% | 405.33 | 100 MG
SDP
Catalog No. 5000343905
Encompass_Preferred
UK122 is a potent and selective urokinase-type plasminogen activator (uPA) inhibitor with an IC50 of 0.2 μM. It exhibits minimal inhibition of tissue-type PA (tPA), plasmin, thrombin, and trypsin (all IC50 >100 μM). This 4-oxazolidinone analogue acts as an anticancer agent by inhibiting cancer cell migration and invasion.
  • Dose-dependently inhibits cell migration
  • Significantly reduces invasiveness of CFPAC-1 cells
  • Does not affect cell growth or cell morphology in CFPAC-1 cells at certain concentrations
  • Appears as a white to yellow solid
  • Recommended for research use only
Catalog No. 50-003-43905 Supplier Medchemexpress LLC Supplier No. HY111056100MG
UK122 is a potent and selective urokinase-type plasminogen activator (uPA) inhibitor with an IC50 of 0.2 μM. It exhibits minimal inhibition of tissue-type PA (tPA), plasmin, thrombin, and trypsin (all IC50 >100 μM). This 4-oxazolidinone analogue acts as an anticancer agent by inhibiting cancer cell migration and invasion.
  • Dose-dependently inhibits cell migration
  • Significantly reduces invasiveness of CFPAC-1 cells
  • Does not affect cell growth or cell morphology in CFPAC-1 cells at certain concentrations
  • Appears as a white to yellow solid
  • Recommended for research use only