Learn More
Biolegend Ultra-LEAF™ Purified Human IgG4 Isotype Control Recombinant
Supplier:  Biolegend 403701
Encompass_Preferred
Ultra-LEAF™ Purified Human IgG4 Isotype Control Recombinant [QA16A15]; Isotype: Human IgG4, κ; Apps: FC; Size: 100 ug
Catalog No. NC1554428