Learn More
Hach Company UL Range CL17sc w/SC4500 Kit
Supplier:  Hach Company 9793400
Encompass
Ultra Low Range CL17sc Colorimetric Chlorine Analyzer with Pressure Regulator Installation Kit and Reagents for Total ChlorineULR CL17sc Chlorine Analyser with Pressure Regulation Installation Kit (9790300)-Total Chlorine Indicator Solution (2263411)-Total Chlorine Buffer Solution (2263511)-DPD Compound (2297255)-User Manual
Catalog No. 50-002-07711
Add to Cart
Request a Quote