Learn More
Hach Company UL Range CL17sc w/SC4500 Kit
Catalog No. 5000207588 Shop All Hach Company Products
Encompass
Ultra Low Range CL17sc Total Chlorine Analyzer with Standpipe Installation Kit-SC4500 Controller and ReagentsULR CL17sc Chlorine Analyser with Standpipe Installation Kit (9790200) SC4500 Controller for 2 digital sensors-Prognosys-5x mA Output-100-240 VAC-without power cord (LXV525 99A11551) Total Chlorine Indicator Solution (2263411) Total Chlorine Buffer Solution (2263511) DPD Compound (2297255) User Manual
Catalog No. 50-002-07588 Supplier Hach Company Supplier No. 9792700
Add to Cart
Add to Cart
Request a Quote
Ultra Low Range CL17sc Total Chlorine Analyzer with Standpipe Installation Kit-SC4500 Controller and ReagentsULR CL17sc Chlorine Analyser with Standpipe Installation Kit (9790200) SC4500 Controller for 2 digital sensors-Prognosys-5x mA Output-100-240 VAC-without power cord (LXV525 99A11551) Total Chlorine Indicator Solution (2263411) Total Chlorine Buffer Solution (2263511) DPD Compound (2297255) User Manual