Learn More
Hach Company UL Range CL17sc w/SC4500 Kit
Catalog No. 5000207589 Shop All Hach Company Products
Encompass
Ultra Low Range CL17sc Total Chlorine Analyzer with Pressure Regulator Installation Kit and SC4500 Controller and ReagentsULR CL17sc Chlorine Analyser with Pressure Regulation Installation Kit (9790300) SC4500 Controller for 2 digital sensors-Prognosys-5x mA Output-100-240 VAC-without power cord (LXV525 99A11551) Total Chlorine Indicator Solution (2263411) Total Chlorine Buffer Solution (2263511) DPD Compound (2297255) User Manual
Catalog No. 50-002-07589 Supplier Hach Company Supplier No. 9792800
Add to Cart
Add to Cart
Request a Quote
Ultra Low Range CL17sc Total Chlorine Analyzer with Pressure Regulator Installation Kit and SC4500 Controller and ReagentsULR CL17sc Chlorine Analyser with Pressure Regulation Installation Kit (9790300) SC4500 Controller for 2 digital sensors-Prognosys-5x mA Output-100-240 VAC-without power cord (LXV525 99A11551) Total Chlorine Indicator Solution (2263411) Total Chlorine Buffer Solution (2263511) DPD Compound (2297255) User Manual