Learn More
Hach Company UL Range CL17sc w/SC4500 Kit
Supplier:  Hach Company 9793300
Encompass
Ultra Low Range CL17sc Colorimetric Chlorine Analyzer with Standpipe Installation Kit and Reagents for Total ChlorineULR CL17sc Chlorine Analyser with Standpipe Installation Kit (9790200)-Total Chlorine Indicator Solution (2263411)-Total Chlorine Buffer Solution (2263511)-DPD Compound (2297255)-User Manual
Catalog No. 50-002-07839
Add to Cart
Request a Quote