Learn More
Sino Biological ULBP2 / N2DL-2 Antibody, Rabbit MAb 100uL
Supplier:  Sino Biological 12143R022100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human ULBP2 / N2DL-2 (rh ULBP2 / N2DL-2; Catalog#12143-H08H; Q9BZM5; Met1-Ser217).
Catalog No. 50-224-7327