Learn More
Discovery Life Sciences Ulcerative Colitis PBMC
SDP
Catalog No. 5000804010
Encompass
PBMCs Peripheral Blood Mononuclear Cells are immune cells isolated from whole blood and viably cryopreserved providing a foundational material for immunology studies cell therapy research and functional immune assays
Catalog No. 50-008-04010 Supplier Discovery Life Sciences Supplier No. 121570
PBMCs Peripheral Blood Mononuclear Cells are immune cells isolated from whole blood and viably cryopreserved providing a foundational material for immunology studies cell therapy research and functional immune assays