Learn More
Biolegend Ultra-LEAF™ Purified anti-c-Myc
Supplier:  Biolegend 626807
Encompass_Preferred
Ultra-LEAF™ Purified anti-c-Myc [9E10]; Isotype: Mouse IgG1, κ; Reactivity: Endogenous Human c-Myc; fusion protein with c-Myc tag (EQKLISEEDL, around 1.2kD) in all species; Apps: WB, ELISA, ICC, IP, IHC-P, Purification; Size: 100 ug
Catalog No. 50-208-6834