Learn More
Biolegend Ultra-LEAF™ Purified anti-CCL15
Supplier:  Biolegend 692704
Encompass_Preferred
Ultra-LEAF™ Purified anti-CCL15 [A15015A]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: Block; Size: 1 mg
Catalog No. 50-208-7614